Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
Brand:  Hirschmann™ 9211380
 View more versions of this product
Product Code. 13270490
  • 75.70 € / Pack of 1000
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Specifications
0.95 mm
Melting point determination
80 mm
1.35 mm
One end closed
Capillary Tube