Learn More
Hirschmann™ Capillary Tubes, Both Ends Open
Excellent for working with small samples in the laboratory. Can be utilized for a wide range of scientific and medical applications.
Brand:  Hirschmann™ 9201512
 View more versions of this product
Product Code. 10623202
  • 87.25 € / Pack of 1000
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Both ends open
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Capillary Tubes, Both Ends Open
1.15 mm
120 mm
4 x 250/Pk.