Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand:  Hirschmann™ 9211075
 View more versions of this product
Product Code. 10673192
  • 96.20 € / Pack of 1000
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Melting Point Determination Tube, One End Closed
0.80 mm
75 mm
10 x 100/Pk.