missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capitol Vial Snappies™ Breast Milk Containers
GREENER_CHOICE

Collect, store and dispense breast milk with Snappies containers.

Brand:  Thermo Scientific™ 10253

177.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15668558

  • 185.00 € / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Snappies breast milk containers are built for purpose to support the nutritional requirements of premature infants in their critical, early months. The containers are offered in 1oz. to 2.3 oz. volumes, to scale with mother's milk production, and feature a unique flip-top design that enables collection, storage and feeding all from the same bottle.

Features:

  • Easy-open tab design
  • Locking hinged lid stays out of the way during use
  • Easy to see milk volume numbers
  • Large base protects container from falling over
  • Fits most standard size nipples used for feeding and threaded breast pumps
  • Available in two convenient sizes: 1 oz. (29.6mL) and 2.3 oz. (70mL)
  • Calibrated in mLs for accurate feeding measurements
  • Space to write mother's name, date and collection time

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Breast Milk Container
2.3 oz.
Flip Top
Space to write mother's name, date, and collection time
Polypropylene
70mL
CV Snappies 70mL Human Milk Container (20 packs of 10)
200/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.