Learn More
Thermo Scientific™ Capitol Vial Split Specimen Urine Collection Kit
Perform sample collection and storage with these capitol vial split foil top specimen collection kits.
206.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15698568
![]() |
Thermo Scientific™
92023 |
Split Foil Top Specimen Collection Kit, 100/Cs |
206.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Capitol Vial Split Specimen Collection Kit complies with SAMHSA and Department of Transportation guidelines for urine drug testing programs and other applications where a second sample is required.
- Easy-to-use—simply peel away the foil top lid, remove the components and provide donor with the collection cup
- After sample collection, 30mL of urine is poured into the first transport vial and 15mL into the second vial, close the hinged lid until it snaps
- Place vial in front pocket of specimen bag and completed requisition in the rear pocket
- Remove the blue release liner on the bag and fold flap over the pockets to seal
Ordering Information:
Custom branding available for foil tops and specimen bags.
Includes:
Beaker (8 oz.) with temperature indicator strip, 2 to 45mL Custody vials, Generic dual-pocket specimen bag, absorbent foil top.
Note:
For testing according to Substance Abuse and Mental Health Services Administration guidelines, refer to SAMSHA instruction for collection and transport requirements.
Specifications
Specimen Collection Kit |