missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capping Reagent, Honeywell Burdick & Jackson™

Click to view available options
:
4X2.5L GLASS BOTTLE
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Spezifikation
Spezifikation
| CAS | 110-86-1, 108-24-7, 109-99-9 |
| CAS Max % | 10.00%, 80.00% |
| Boiling Point | 66°C.The physical data is that of the main component. |
| Quantity | 4 x 2.5 L |
| Flash Point | -15°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid, clear |
| Purity Grade Notes | Capping reagents |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur