Learn More
Description
Used to formulate CAPS buffer, a zwitterionic buffer useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electro transfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5. and minimal reactivity with enzymes or proteins, minimal salt effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used to formulate CAPS buffer, a zwitterionic buffer useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electro transfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI>8.5. and minimal reactivity with enzymes or proteins, minimal salt effects.
Solubility
Highly water soluble (100 g/L) at 20°C, and methanol (1%).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Specifications
Specifications
Melting Point | 324°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 2835588 |
Merck Index | 14,1767 |
Solubility Information | Highly water soluble (100g/L) at 20°C,and methanol (1%). |
Formula Weight | 221.32 |
Percent Purity | 99% |
Chemical Name or Material | CAPS |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0460881 | Certificate of Analysis | 04/04/2024 | A17037.22,A17037.14,A17037.36 |
A0456601 | Certificate of Analysis | 22/02/2024 | A17037.22,A17037.14,A17037.36 |
N05K042 | Certificate of Analysis | 14/02/2024 | A17037.22,A17037.14,A17037.36 |
A0446305 | Certificate of Analysis | 18/11/2022 | A17037.22,A17037.14,A17037.36 |
W01I019 | Certificate of Analysis | 21/09/2022 | A17037.22,A17037.14,A17037.36 |
Safety and Handling

- CAPS
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : CAPS
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.