missing translation for 'onlineSavingsMsg'
Learn More

CAPS (Fine White Crystals), Fisher BioReagents

44.80 € - 170.00 €

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym 3-Cyclohexylamino-1-propanesulfonic Acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071453
View Documents
Fisher BioReagents
BP321-100
100 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10081453
View Documents
Fisher BioReagents
BP321-500
500 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This biological buffer has a usable pH range of 9.7 to 11.1.
Specifications

Chemical Identifiers

1135-40-6
221.32
PJWWRFATQTVXHA-UHFFFAOYSA-N
70815
OS(=O)(=O)CCCNC1CCCCC1
C9H19NO3S
MFCD00003837
3-Cyclohexylamino-1-propanesulfonic Acid
3-(cyclohexylamino)propane-1-sulfonic acid

Specifications

1135-40-6
White
Powder or Solid
Pass Test
C6H11NH(CH2)3SO3H
15, 1769
3-Cyclohexylamino-1-propanesulfonic Acid
OS(=O)(=O)CCCNC1CCCCC1
221.32
221.32
CAPS
324°C
3.0 to 7.0
≥98 %
C9H19NO3S
MFCD00003837
Negligible
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
70815
≥98%
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : RT

Product Identifier
  • CAPS

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.