missing translation for 'onlineSavingsMsg'
Learn More
Capsazepine, Tocris Bioscience™ Product Code.: 30001093

Capsazepine, Tocris Bioscience™

Product Code. 30001093
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001093

Brand: Tocris Bioscience 0464/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vanilloid receptor antagonist; also activator of ENaCδ

Selective vanilloid receptor antagonist (Ki = 3.2 μM). Inhibits carrageenan inflammation-induced hyperalgesic responses in the rat. Also activates amiloride-sensitive epithelial Na+ channel ENaCδ.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138977-28-3
Molecular Formula C19H21ClN2O2S
Molecular Weight (g/mol) 376.899
InChI Key DRCMAZOSEIMCHM-UHFFFAOYSA-N
Synonym capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
PubChem CID 2733484
ChEBI CHEBI:70773
IUPAC Name N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
SMILES C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O

Specifications

Quantity 50 mg
Formula Weight 376.9
Percent Purity >99%
Chemical Name or Material Capsazepine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Capsazepine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.