Learn More
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ H402 Version 03 Filtration Columns are for storing organic vapors and acids
Brand:  Erlab™ G0103/BE
 View more versions of this product
Product Code. 12157720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Horizontal filtration column
  • HEPA H14 filter
  • Handlings in an ultra clean environment
  • Serial prefilter(optimizes the HEPA H14 filter lifetime)
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
Specifications
Specifications
Filtration Column
BE Filter
For storing organic vapors and acids
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products