Learn More
Erlab™ Captair™ Filtration Column for Fume Hoods
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Filtration Columns for Fume Hoods are configured according to the protection needs of the hood’s specific application.
Brand:  Erlab™ G0102/AS
 View more versions of this product
Product Code. 12107720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Modular filtration column
  • Handlings in an ultra-clean environment
  • Serial prefilter (optimizes the HEPA H14 filter lifetime)
  • Particulate HEPA H14 filtration
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
  • Equipped with a HEPA H14 filter, guarantees 99.995% filtration efficiency for particles larger than 0.1μm (according to the MPPS method set forth in the EN 1822-1 standard)
  • Ultra-clean air entering the enclosure meets ISO class 5* (EN ISO 14-644 standard) requirements, which corresponds to American class 100 (i.e., less than 100 particles per cubic foot > 0.5 μm) and to class A and B of the GMP guide published by the European Union for the pharmaceutical industry
  • Items located into the enclosure are therefore protected from any external contaminants
  • HEPA filtered enclosure
Applications:
  • Non-pathogenic cell cultures
  • In-vitro cultures
  • Microbiology (Non-pathogenic)
  • Homeopathic preparations in pharmacies
  • Electronics
  • Optics, etc.
Specifications
Specifications
Securifilter AS Filter
02 Sampling Port, Fan Failure Alarm, Securifilter AS Filter
Horizontal Filtration Column
For storage with organic vapors mainly for standard vertical safety cabinets
267 mm
570 mm
390 mm