Learn More
Erlab™ Captair™ Filtration Column for Fume Hoods
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Filtration Columns for Fume Hoods are configured according to the protection needs of the hood’s specific application.
Brand:  Erlab™ G0201/BE
 View more versions of this product
Product Code. 12157730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Modular filtration column
  • Handlings in an ultra-clean environment
  • Serial prefilter (optimizes the HEPA H14 filter lifetime)
  • Particulate HEPA H14 filtration
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
  • Equipped with a HEPA H14 filter, guarantees 99.995% filtration efficiency for particles larger than 0.1μm (according to the MPPS method set forth in the EN 1822-1 standard)
  • Ultra-clean air entering the enclosure meets ISO class 5* (EN ISO 14-644 standard) requirements, which corresponds to American class 100 (i.e., less than 100 particles per cubic foot > 0.5 μm) and to class A and B of the GMP guide published by the European Union for the pharmaceutical industry
  • Items located into the enclosure are therefore protected from any external contaminants
  • HEPA filtered enclosure
Applications:
  • Non-pathogenic cell cultures
  • In-vitro cultures
  • Microbiology (Non-pathogenic)
  • Homeopathic preparations in pharmacies
  • Electronics
  • Optics, etc.
Specifications
Specifications
BE
01 Sampling Port, Securifilter BE Filter
Vertical Filtration Column
For storing organic vapors and Acids for under bench safety cabinets
587 mm
537 mm
200 mm