missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair™ Smart 481 Ductless Fume Hood

Designed with advanced carbon filtration to minimize chemical exposure.

Specifications

Product Type Ductless Filter Hood
View More Specs

Products 4
Product Code Brand Filter Type For Use With (Application) Includes Price Quantity & Availability  
Product Code Brand Filter Type For Use With (Application) Includes Price Quantity & Availability  
15594360
View Documents Promotion Details
Erlab™
A62B/AS
Class 1, Type 2C, Liquid Chemical Handling For organic vapors 2 carbon filter N/A
N/A
15564360
View Documents Promotion Details
Erlab™
A62A/BE
Class 2, Type 1C, Carbon Filters, Liquid Chemical Handling For organic vapor, acid 2 carbon filter N/A
N/A
15514380
View Documents Promotion Details
Erlab™
A62E/HP
HEPA, Type 1P For organic vapors One filter N/A
N/A
15584370
View Documents Promotion Details
Erlab™
A62D/BE
Type 1P, Type 2C+, GF4.BE Carbon filter Acid/Organic Vapours N/A
N/A
Description

Description

  • Sensors to detect filter breakthrough of solvents, acids or formaldehyde
  • Safety back up filter in case of main filter saturation
  • Continuous monitoring of air face velocity

Erlab Safety Program

  • Application analysis and validation, usage certification, filter change reminders
  • Connect with eGuard™ for real-time status, notifications and alerts
  • Smart-Light feature for lighting up the hood

Flexibility

  • Configurable filtration column
  • No ductwork needed

Environment

  • No dangerous chemicals are released into atmosphere
  • Low energy consumption
Specifications

Specifications

Ductless Filter Hood
Videos
SDS
Documents

Documents

Product Certifications