missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto Q, 25mL

Designed to meet the demands of modern large-scale manufacturers. Cytiva Capto Q, 25ml is a strong anion exchange BioProcess medium, for fast, efficient, and cost-effective capture and intermediate protein purification of quaternary amine ligands.

Brand:  Cytiva 17-5316-10


Product Code. 10393490

  • 90.00 € / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Capto Q is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto Q raises the productivity in industrial downstream processes. Capto Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
  • Strong anion exchanger for high-productivity capture and intermediate purification when high volume throughput is essential
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations
  • Meets industrial demands: security of supply, technical and regulatory support
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Capto Q, 25 mL
Designed to meet the demands of modern, large-scale manufacturers Capto Q’s shorter processing times can reduce exposure of the target protein.
4°C to 30°C in 20% Ethanol
2 to 12
Highly cross-linked agarose with dextran surface extender
Quaternary amine
90 μm, d50v is the median particle size of the cumulative volume distribution.
25 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Capto Q, 25 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers