Learn More
Cytiva Capto™ S Chromatography Media
Designed for capture and intermediate purification of proteins from large feed volumes. Cytiva CaptoTM S Chromatography Media is a strong cation exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers.
Brand: Cytiva 17-5441-01
334.40 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- Rigid, high flow agarose matrix modified with dextran surface extenders and strong sulfonate cation exchanger
- Combination of high volume throughput and dynamic binding capacity at high flow for raise in productivity
- Withstands effective CIP procedure and is chemically stable
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications
100 mL | |
For Fast, Efficient Capture and Intermediate Protein PurifiCatIon | |
Capto S Media | |
90 μm | |
20 cm | |
Strong CatIon Exchanger | |
Highly Cross-Linked Agarose with Dextran surface extender | |
20% Ethanol, 0.2 M Sodium Acetate |
1 M Acetic Acid, 1 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol | |
4°C to 30°C | |
700 cm/Hoμr | |
7.87 in. | |
0.11 to 0.14 mM Na+/mL | |
300 kPa | |
3 to 14 | |
Sulphonate Group |
Safety and Handling
- Capto S, 100 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.