missing translation for 'onlineSavingsMsg'
Learn More

Carbadox, 98+%, Thermo Scientific Chemicals

Product Code. 15463079
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6804-07-5
MDL Number MFCD00057293

Specifications

Melting Point 239°C to 240°C
Color Yellow
Flash Point 18°C (64°F)
Molecular Formula C13H10BrNO
Quantity 25 g
UN Number UN1325
Merck Index 14,1780
Synonym 4-bromobenzanilide, n-phenyl 4-bromobenzamide, benzamide, 4-bromo-n-phenyl, 4-bromo-n-phenyl-benzamide, 4-bromo-n-phenyl-menzamide, maybridge1_004278, n1-phenyl-4-bromobenzamide, 4-bromophenyl-n-benzamide
Solubility Information Soluble in 1N NaOH (50mg/ml),and water (partly).
InChI Key QVTGXXQAVWWJRO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Br
IUPAC Name 4-bromo-N-phenylbenzamide
Molecular Weight (g/mol) 276.13
PubChem CID 690966
Formula Weight 262.22
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Carbadox
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.