missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine chloride, 99%, Thermo Scientific Chemicals

Product Code. 11390622
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-carbamoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

Melting Point 200°C to 204°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula H2NCO2CH2CH2N(CH3)3Cl
Merck Index 15,1781
Solubility Information Solubility in water: 1g/mL. Other solubilities: 1g/50 mL alcohol,1g/10 mL methanol,practically insoluble in chloroform and ether
Formula Weight 182.65
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 2.0% max. (105°C, 2 hrs)
Packaging Glass bottle
Chemical Name or Material Carbamylcholine chloride
Show More Show Less
compliance-icons
Product Identifier
  • Carbamylcholine chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'