missing translation for 'onlineSavingsMsg'
Learn More

Carbenoxolone disodium, Tocris Bioscience™

Product Code. 30001447
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Gap junction blocker. Also inhibitor of 11 β-hydroxysteroid dehydrogenase

Glucocorticoid that inhibits 11β-hydroxysteroid dehydrogenase (11-HSD) and blocks gap junction communication.

Specifications

CAS 7421-40-1
Molecular Formula C34H48Na2O7
MDL Number MFCD00079043
Quantity 50 mg
Synonym carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate
InChI Key BQENDLAVTKRQMS-SBBGFIFASA-L
SMILES [Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O
IUPAC Name disodium (2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-[(3-carboxylatopropanoyl)oxy]-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylate
Molecular Weight (g/mol) 614.73
PubChem CID 133687779
Formula Weight 614.72
Percent Purity >98%
Chemical Name or Material Carbenoxolone disodium
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.