missing translation for 'onlineSavingsMsg'
Learn More

Carbimazole, 98%, Thermo Scientific™

Product Code. 15500584 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15500584

Brand: Thermo Scientific Acros 458610250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22232-54-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
InChI Key CFOYWRHIYXMDOT-UHFFFAOYSA-N
Synonym carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
PubChem CID 31072
ChEBI CHEBI:617099
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C1=S)C

Specifications

Melting Point 122.0°C to 124.0°C
Loss on Drying 0.5% max. (24 h) (P2O5)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: 2g/L ( 20°C ). Other solubilities: soluble in acetone and ethanol
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material Carbimazole
compliance-icons
Product Identifier
  • Carbimazole
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.