Learn More
Thermo Scientific Chemicals Carbol Fuchsin, pure
Chemical Identifiers
CAS | 4197-24-4 |
---|---|
Molecular Formula | C21H22ClN3 |
Molecular Weight (g/mol) | 351.878 |
MDL Number | MFCD00143923 |
InChI Key | HZLHRDBTVSZCBS-GHTYLULLSA-N |
PubChem CID | 91997463 |
IUPAC Name | 4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride |
SMILES | [H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636972
|
Thermo Scientific Chemicals
195790050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10134351
|
Thermo Scientific Chemicals
195790250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10723285
|
Thermo Scientific Chemicals
195791000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4197-24-4 | |
351.878 | |
HZLHRDBTVSZCBS-GHTYLULLSA-N | |
4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride |
C21H22ClN3 | |
MFCD00143923 | |
91997463 | |
[H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-] |
Specifications
4197-24-4 | |
MFCD00143923 | |
Authentic | |
4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride | |
91997463 | |
Pure | |
Complies | |
Crystalline Powder |
C21H22ClN3 | |
HZLHRDBTVSZCBS-GHTYLULLSA-N | |
[H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-] | |
351.878 | |
(in water) about 542nm | |
(for microscopy) passes | |
Green | |
Carbol Fuchsin, A mixture of Basic Fuchsin and phenol. |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing genetic defects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical attention/advice.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'dotInformation' : DOT Class 6.1
missing translation for 'einecsNumber' : 224-086-6
RUO – Research Use Only