Learn More
Brannan™ Carbon Dioxide Detector
The Brannan carbon dioxide detector measures Carbon Dioxide (CO2), air temperature and humidity. Comes complete with certificate of calibration.
Brand:  Brannan™ 38/920/0
Product Code. 17435313
  • 243.00 € / Each
Explore more special offers
Description
Description
  • Measures ventilation in enclosed spaces such as classrooms and workplaces
  • Indoor air quality displayed in ppm with colour coded good, normal and poor indicators
  • This device adopts a high-precision non-dispersive infrared CO2 sensor that detects CO2 accurately and sensitively
  • This product uses a high-capacity rechargeable lithium battery inside, making it ideal to move around a space
  • To preserve electricity, the device powers off after 30 minutes of inactivity
  • A USB charging cable is provided allowing the user to recharge the device using either a PC or mains plug
Specifications
Specifications
CE/RoHS/WEEE
Carbon Dioxide
0.5302 lb.
0 to 9999 ppm, -20°C to +70°C, 0 to 100% RH
Non-dispersive infrared CO2 sensor
0.2405 kg