missing translation for 'onlineSavingsMsg'
Learn More

Carbon Dioxide Standard (CO2) 6 g/L, Reagecon

One of a series of standards used to calibrate and control the Corning 965D Carbon Dioxide Analyzer.

Brand:  Reagecon SC026C

Product Code. 11728465

  • 203.00 € / 100 ampoules
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For calibrating carbon dioxide analyzer.
  • Useful for determining carbon dioxide content in carbonated beverages.
  • Uses raw materials of very high purity and specification.
  • Tested titremetrically using Reagecon's in-house method TPATB against hydrochloric acid, analytical volumetric standard solution.
  • Standard is directly traceable to NIST Standard Reference Material 723e - tris(hydroxymethyl)aminomethane.
  • A detailed Certificate of Analysis is issued with each batch manufactured.
Specifications

Specifications

Carbon Dioxide Analysis
100 x 2 mL, 6 g/L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon Dioxide Standard (CO2) 6 g/L, Reagecon > 100 x 2 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.