missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, ≥99.9%, Reagent Grade, Honeywell™

Product Code. 15665490
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, purified by redistillation,≥99.9%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-15-0
Density 1.266g/mL at 25°C
Quantity 100 mL
Explosion Limit 0.5
Flash Point −30°C (−22°F)
Ignition Point 212°F
Packaging Glass Bottle
Molecular Formula CS2
MDL Number MFCD00011321
UN Number UN1131
Beilstein 1098293
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES C(=S)=S
IUPAC Name methanedithione
Molecular Weight (g/mol) 76.131
PubChem CID 6348
ChEBI CHEBI:23012
Formula Weight 76.14g/mol
Percent Purity ≥99.9%
Grade ReagentPlus™
Chemical Name or Material Carbon disulfide, Purified by redistillation
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'