missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, 99.9%

Product Code. 15481257
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 15481257

missing translation for 'mfr': Thermo Scientific Alfa Aesar 032472.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-15-0 | CS2 | 76.131 g/mol

Carbon disulfide is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point °C to 112°C
Density 1.2632
Boiling Point 46°C
Quantity 250 mL
Flash Point -30°C (-22°F)
Physical Form Liquid
Refractive Index 1.627
Assay Percent Range 99.9+%
Solubility Information Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Chemical Name or Material Carbon disulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbon disulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Carbon disulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.