missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Carbon powder, activated, Norit™ D10

Product Code. 15417687
Click to view available options
2000g
This item is not returnable. View return policy
This item is not returnable. View return policy
Applications
Carbon powder is used as such in purifying metals, in electrodes, electrical devices and steel.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-44-0
Color Black
Molecular Formula C
MDL Number MFCD00133992
Quantity 2000 g
UN Number UN1362
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Physical Form Powder
Chemical Name or Material Carbon powder, Activated, Norit™ D10
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.