missing translation for 'onlineSavingsMsg'
Learn More

Carbon powder, activated, Norit GSX, steam activated, acid washed, Thermo Scientific Chemicals

30.45 € - 227.13 €

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312677
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11860.22
100 g
30.45 €
100g
Please to purchase this item. Need a web account? Register with us today!
11322677
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11860.36
500 g
72.56 €
500g
Please to purchase this item. Need a web account? Register with us today!
11332677
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11860.0P
2000 g
227.13 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Decolorizing carbon. High purity grade with high internal surface area, suitable for decolorization, purification and catalyst carrier applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-44-0
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7440-44-0
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
C
UN1362
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Carbon powder, activated, Norit GSX, steam activated, acid washed
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities; may catch fire.

P235+P410-P280

H252

missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: CARBON, ACTIVATED

missing translation for 'einecsNumber' : 231-153-3

missing translation for 'rtecsNumber' : FF5250100

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only