missing translation for 'onlineSavingsMsg'
Learn More

Carbon sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15463735
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Used as Magnetron sputtering source. The carbon sputtering target was replaced after the de- positions of 20-30 films with several thousand A in thick- ness in mass-separated negative-ion-beam deposition system. Carbon films can also be deposited by magnetron sputtering in which a magnetically confined plasma torus (race track) acts as a distributed source of ions that impact a carbon sputtering target over a significant part of its area. C -ions were produced from a highly pure carbon-sputtering target by cesium ion sputtering in a neutral and ionized alkaline bombardment-type heavy negative ion source. CN negative ions of 0.88 mA were safely obtained by using a carbon sputtering target and nitrogen gas instead of cyanogen for ionization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
MDL Number MFCD00133992
Solubility Information Insoluble in water.
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Carbon sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.