Learn More
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 21
Large version of No. 10 blade with a curved, cutting edge and a flat, unsharpened back edge.
Brand:  Swann-Morton™ 3007
Product Code. 15813252
  • 21.40 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Large version of No. 10 blade with a curved, cutting edge and a flat, unsharpened back edge.
  • Large version of No. 10 blade.
  • Curved cutting edge.
  • Flat, unsharpened back edge.
  • Larger than the No. 20.
  • Smaller than the No. 22.
  • Fits handles 4, 4L, 4 graduated, and 6B.
Specifications
Specifications
Carbon Steel
Non-sterile
100/Box
Individually Packaged