Learn More
Swann-Morton™ Carbon Steel Postmortem Scalpel Blades
Use Swann Morton™ Carbon Steel Postmortem Scalpel Blades to facilitate pathology operations.
Brand:  Swann-Morton™ 2551
 View more versions of this product
Product Code. 11360905
  • 49.55 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Added safety feature of a 'bull nose tip' to help offset the occurrence of accidental sharp related injuries
  • Heavy duty carbon steel blade
  • Metal handle with screw-type fitting
  • PM-40
.
Specifications
Specifications
Carbon Steel
# PM40
Sharp
Sharp
SCA-710-K Swann-Morton
10/Pk.