Learn More
Swann-Morton™ Carbon Steel Postmortem Scalpel Blades
Use Swann Morton™ Carbon Steel Postmortem Scalpel Blades to facilitate pathology operations.
Brand:  Swann-Morton™ 2552
 View more versions of this product
Product Code. 12952555
  • 42.75 € / Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Added safety feature of a 'bull nose tip' to help offset the occurrence of accidental sharp related injuries
  • Blunted
  • PM-40
Specifications
Specifications
Carbon Steel
For Postmortem procedures with the added safety feature of a 'bull nose tip' to help offset the occurrence of Accidental Sharp related Injuries
The PM40 Stainless, PM40 Standard or PM40 Plated Handle
Blunted Knife
# PM40 B