Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
Brand:  Swann-Morton™ 0201
 View more versions of this product
Product Code. 11728363
  • 25.20 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Carbon steel to BS EN 27740
  • Sterile
Specifications
Specifications
Carbon Steel
# 10
Sterile
10/Pk.
Scalpel Blade
The # 10 fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
CE compliant 93/42/EC