missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.6

78.89 € - 122.61 €

Specifications

Concentration 0.5 M buffer soln.
pH Range 9.6
Type Carbonate buffer soln.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15414949
View Documents Promotion Details
Thermo Scientific Chemicals
J62610.AK
250 mL
78.89 €
250mL
In Stock
to see stock.
Add to basket
 
15424949
View Documents Promotion Details
Thermo Scientific Chemicals
J62610.AP
500 mL
122.61 €
500mL
In Stock
to see stock.
Add to basket
 
Description

Description

Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

Solubility
It is soluble in water.

Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.
Specifications

Specifications

0.5 M buffer soln.
9.6
Carbonate buffer soln.
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only