Learn More
Invitrogen™ Carbonic Anhydrase I Monoclonal Antibody (K16257_10H9), Invitrogen™
Mouse Monoclonal Antibody
438.00 € - 658.00 €
Specifications
| Antigen | Carbonic Anhydrase I |
|---|---|
| Clone | K16257_10H9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200869
|
Invitrogen™
MA555654 |
50 μL |
438.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30200974
|
Invitrogen™
MA556034 |
200 μL |
658.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KNGPEQWSKL YPIANGNNQS PVDIKTSETK HDTSLKPISV SYNPATAKEI INVGHSFHVN FEDNDNRSVL KGGPFSDSYR LFQFHFHWGS TNEHGSEHTV DGVKYSAELH VAHWNSAKYS SLAEAASKAD GLAVIGVLMK VGEANPKLQK VLDALQAIKT KGKRAPFTNF DPSTLLPSSL DFWTYPGSLT HPPLYESVTW IICKESISVS SEQLAQFRSL LSNVEGDNAV PMQHNNRPTQ PLKGRTVRAS
Carbonic Anhydrase I Monoclonal antibody specifically detects Carbonic Anhydrase I in Human, Rat samples. It is validated for Western BlotSpecifications
| Carbonic Anhydrase I | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AW555628; CA I; CA1; ca1 {ECO:0000250; CAB; CA-I; CA-I {ECO:0000250; Car1; Car-1; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250; carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250; Carbonic anhydrase B; carbonic anhydrase I; carbonic anhydrase I {ECO:0000250; epididymis secretory protein Li 11; HEL-S-11; UniProtKB:P00915} | |
| Recombinant human CA1 (immunogen range is 11-260). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CA1 |
| K16257_10H9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| B0BNN3, P00915 | |
| 310218, 759 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.