Learn More
Carbonic Anhydrase I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA597527
 View more versions of this product
Product Code. 16510362
  • 423.00 € / 100µg
Explore more special offers
Description
Description
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase I
Polyclonal
Unconjugated
CA1
AW555628, CA I, CA1, ca1 {ECO:0000250, CAB, CA-I, CA-I {ECO:0000250, Car1, Car-1, Carbonate dehydratase I, carbonate dehydratase I {ECO:0000250, carbonic anhydrase 1, carbonic anhydrase 1 {ECO:0000250, Carbonic anhydrase B, carbonic anhydrase I, carbonic anhydrase I {ECO:0000250, epididymis secretory protein Li 11, HEL-S-11, UniProtKB:P00915}
Rabbit
Protein G
RUO
12346, 759
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot
3.3 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4
P00915, P13634
CA1, Car1
Recombinant Human Carbonic anhydrase 1 protein (2-261AA)
100 μg
Primary
Human, Mouse