missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase III Monoclonal Antibody (5G8B6), Invitrogen™

Mouse Monoclonal Antibody
258.00 € - 449.00 €
Specifications
| Antigen | Carbonic Anhydrase III |
|---|---|
| Clone | 5G8B6 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17779853
|
Invitrogen™
MA543509 |
50 μL |
258.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17769543
![]() |
Invitrogen™
MA542991 |
100 μL |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human Carbonic Anhydrase III/CA3
Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.Specifications
| Carbonic Anhydrase III | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP | |
| Ca3 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 5G8B6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07451 | |
| 761 | |
| Recombinant Human Carbonic Anhydrase III/CA3 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Ca3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title