missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase X Monoclonal Antibody (7H8F12E6), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

258.00 € - 449.00 €

Specifications

Antigen Carbonic Anhydrase X
Clone 7H8F12E6
Concentration 1 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17759853
View Documents
Invitrogen™
MA543507
50 μL
258.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17719553
GREENER_CHOICE
View Documents
Invitrogen™
MA542996
100 μL
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specificity: Human CA10/CARPX

This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

Carbonic Anhydrase X
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative
2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076
CA10
IgG1
Protein A
Antibody
7H8F12E6
ELISA
Unconjugated
Mouse
Human
Q9NS85
56934
Recombinant Human CA10 protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CA10
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.