missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Description
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.
Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.
Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Specifications
Melting Point | ∽175°C (decomposition) |
Odor | Odorless |
Quantity | 100 mg |
UN Number | UN3439 |
Beilstein | 1842102 |
Solubility Information | Insoluble in water. Soluble in DMSO (5mg/ml),ethanol (1mg/ml) and methanol (10mg/ml). |
Formula Weight | 204.62 |
Percent Purity | 98% |
Chemical Name or Material | Carbonyl cyanide 3-chlorophenylhydrazone |
Safety and Handling
Product Identifier
- Carbonyl cyanide 3-chlorophenylhydrazone
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Carbonyl cyanide 3-chlorophenylhydrazone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction