missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pohli™ Cargo Card Box
Cargo card box for Pohli SUPER FSP. This box is obligatory for the UN approval of the SUPER FSP combination packaging being valid (6PH1/X2.2/250).
Brand: Pohli™ 23013901
Alert:
The carton overpack (Cargo Box) AND also the styrofoam box (Super FSP) have to be labeled and marked by the filler respectively by the sender with all necessary information and symbols according to the international rules and requirements for transportation of dangerous goods.
Description
- Brown cargo box suitable for 25L Super FSP (03048176)
- With flaps and cutting
- With pull strip as tamper-evident packaging
- With Pohli printing, including label field for your product information
- Dimensions (L x W x H): 448 x 448 x 688mm
- 1.700 g/piece
Specifications
SUPER FSP packaging 03048176 | |
448 x 448 x 688 mm | |
Cardboard |
Brown | |
1.7 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction