Learn More
Carl Zeiss™ Invertoskop™ Universal Digital Camera Adapter
Record microscopic findings digitally with this camera adapter. Carl Zeiss™ Invertoskop™ Universal Digital Camera Adapter fits onto a number of Axio, Stemi, and Stereo microscope lines.
Brand:  Carl Zeiss™ 426126-0000-000
Product Code. 10404425
  • 442.00 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Interface connection for digital camera
  • Suitable for a number of Axio, Stemi, and Stereo microscope lines
Specifications
Specifications
Universal Digital Camera Adapter
d30 M37/52x0.75
Microscope, digital camera
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Carl Zeiss™ Invertoskop™ Universal Digital Camera Adapter > Type: d30 M37/52x0.75