Learn More
Heidolph Instruments Carousel Attachment
Spare attachment for 12 vessels
Brand:  Heidolph Instruments 549-03000-00
Product Code. 10129691
  • 97.40 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Attachment for 12 vessels with diameters from 16 to 32 mm
Specifications
Specifications
Test Tubes
12