Learn More
Fisherbrand™ Universal Carousel Pipette Stand
Holds up to 8 single channel, 4 multi channel, or a combination of both pipettes. Fisherbrand™ Universal Carousel Pipette Stand will securely hold virtually every known brand of pipette in the marketplace making it truly universal.
Brand:  Fisherbrand™ HS120480
121.82 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15650785
  • 160.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Ideal for labs that do not wish to be tied into a specific brand of pipette, or already have multiple brands in use
  • Weighted base allows for easy removal or replacement of pipettes, with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
  • Made of ABS (Acrylonitrile Butadiene Styrene)
Specifications
Specifications
Pipettes (8 single-channel pipettes, 4 multichannel pipettes, or a combination of both)
8
408 mm
ABS
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Universal Carousel Pipette Stand >