missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Carrier RNA

Ambion™ Carrier RNA (1 mg/mL) is a component of all the MagMAX™ Isolation Kits that may be purchased separately.

Brand:  Applied Biosystems™ 4382878

149.50 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 30057787

  • 230.00 € / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Provided in one tube of 500 μL.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store solution at –20°C.
Ambion™
500 μL
Carrier for Nucleic Acid Precipitation
Carrier RNA
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2783372 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4382878
Lot Number2783371 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4382878
Lot Number2787754 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4382878
Lot Number2823356 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4382878
Lot Number2823352 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4382878
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ Carrier RNA > 500 μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.