Learn More
Veolia Water Systems™ Cartidges
Robust and reliable water purification cartridges ideal for removing ionic contaminants to achieve specified water quality.
Brand:  Veolia Water Systems™ CODILC0208
 View more versions of this product
Product Code. 13115980
  • 613.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Water purification cartridge
  • Removes ionic contaminants to achieve specified water quality
  • Compatible with PURELAB Flex
Specifications
Specifications
LC208