missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Cartridge

Gas cartridge, with safety valve.

Brand:  Usbeck Laborgeraete™ 1430


Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10447421

  • 6.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • With safety valve
  • Provides approx. 3 hours use
  • EN 417: 2012
  • 70 % butane; 30 % propane
  • Valve catridge type 1430
  • Weight: 220 g
  • Height: 92 mm
  • Diameter: 104 mm
Specifications

Specifications

Gas Cartridge
400 mL
1 Ea.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gaskartusche / Flüssiggasgemisch

Signal Word
  • Danger

Hazard Category
  • Flammable gas.

Hazard Statement
  • H220-Extremely flammable gas.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Usbeck Laborgeraete™ Cartridge > Size: 400mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.