Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Brand: Thermo Scientific™ 09.1006
575.94 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:- MicroPure™
- GenPure™
- GenPure™ Pro
- GenPure™ xCAD
- Smart2Pure™
- LabTower™ EDI
- Pacific™ TII
Specifications
Ultrapure polisher cartridge | |
MicroPure | |
Cartridge, Ultrapure Polisher |