Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Brand:  Thermo Scientific™ 09.4011
276.86 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
 View more versions of this product
Product Code. 11397151
  • 305.52 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High purity water cartridge
Pacific TII
Cartridge, High-purity Water