missing translation for 'onlineSavingsMsg'
Learn More

(-)-Carveol, mixture of isomers, 97%, Thermo Scientific Chemicals

Product Code. 11409077
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a terpene that inhibits mammary carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a terpene that inhibits mammary carcinogenesis.

Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-48-9
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00869995,MFCD00062993
InChI Key BAVONGHXFVOKBV-UHFFFAOYNA-N
Synonym carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
PubChem CID 7438
ChEBI CHEBI:23046
IUPAC Name 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
SMILES CC(=C)C1CC=C(C)C(O)C1

Specifications

Density 0.96
Boiling Point 227°C
Flash Point 98°C (209°F)
Refractive Index 1.496
Quantity 250 g
Beilstein 6269416
Solubility Information Partly soluble in water. Soluble in alcohol,ethanol and DMSO.
Formula Weight 152.24
Percent Purity 97%
Chemical Name or Material (-)-Carveol, mixture of isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'