missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh

Used as cheese, food additive and binder for safety matches.

Brand:  MP Biomedicals™ 0290452080

 View more versions of this product

Product Code. 11432490

  • 4835.00 € / 100lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphoprotein
  • Extracted from bovine milk
  • Used as food source
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Casein
Powder
100 lb.
Yellow
Used as cheese, food additive and binder for safety matches
Store at room temperature (15°C to 30°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh > Quantity: 100lb

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.