missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
281.00 € - 284.00 €
Specifications
Closure Included | Yes |
---|---|
Packaging Type | Loose Packed (Lids) |
Quantity | 1000/Cs. |
Pore Style | Small Squares |
Closure Type | Separate Lid, Front Hinge |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
16324126
|
Epredia™
B851738BE |
Beige |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16334126
|
Epredia™
B851738BL |
Blue |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16344126
|
Epredia™
B851738GN |
Green |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16354126
|
Epredia™
B851738GY |
Gray |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16364126
|
Epredia™
B851738LA |
Lavender |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16374126
|
Epredia™
B851738LG |
Light Green |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16384126
|
Epredia™
B851738LI |
Lilac |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16394126
|
Epredia™
B851738OR |
Orange |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16304136
|
Epredia™
B851738PK |
Pink |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16314136
|
Epredia™
B851738TN |
Tan |
284.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16324136
|
Epredia™
B851738WH |
White |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16334136
|
Epredia™
B851738YW |
Yellow |
281.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
1000/Cs. | |
Separate Lid, Front Hinge | |
Cassette II with Color-Matched Lids |
Loose Packed (Lids) | |
Small Squares | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title