missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Cassette II Slotted Tissue Cassettes

For routine tissue samples, these loose-packed cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties

227.00 € - 240.00 €

Specifications

Closure Included Yes
Quantity 1000/Cs.
Pore Style Slots
Closure Type Separate Lid, Front Hinge
Product Type Slotted Cassette
View More Specs

Products 11
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16344136
View Documents
Epredia™
B851739BE
Beige
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16354136
View Documents
Epredia™
B851739BL
Blue N/A N/A N/A
16364136
View Documents
Epredia™
B851739GN
Green
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16384136
View Documents
Epredia™
B851739LA
Lavender
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16394136
View Documents
Epredia™
B851739LG
Light Green
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16304146
View Documents
Epredia™
B851739LI
Lilac
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16314146
View Documents
Epredia™
B851739OR
Orange
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16324146
View Documents
Epredia™
B851739PK
Pink
240.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16334146
View Documents
Epredia™
B851739TN
Tan
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16344146
View Documents
Epredia™
B851739WH
White
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16354146
View Documents
Epredia™
B851739YW
Yellow
227.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Slots
Slotted Cassette
1000/Cs.
Separate Lid, Front Hinge
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Cassette II Slotted Tissue Cassettes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.